MW 569.7 Da, Purity >90%. Fluorescent membrane potential indicator. Selectively stains electrically excitable membranes. Stains Ca+-dependently in buffered iso-osmotic sucrose and Ca+-independently at high ionic strength.
The product is subject to the following: Abcam Restricted Use Statement