missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 43-113 Aramid Fiber Flame Resistant Gloves

Catalog No. p-7111645
Change view
Click to view available options
Size:
10
9
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19010288 9
19010289 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19010288 Supplier Ansell Supplier No. 43113090
Only null left

Gloves are resistant to contact, convective, and radiant heat. Designed for intermittent handling of hot, dry objects up to 350°C.

  • Aramid Fiber shell provides resistance to cuts and heat
  • Designed for intermittent handling of dry, hot objects up to 350°C
  • Flame resistant design will not melt, drip, or propogate a flame
  • Gloves are resistant to contact, convective, and radiant heat
  • Long knitwrist cuff extends protection to the wrist

Specifications

ANSI Abrasion Rating ANSI 3
Color Yellow
Cut Resistance Rating ANSI A5
Material Aramid Fiber
Size 9
Temperature (English) Max. 660°F
Temperature (Metric) Max 350°C
For Use With (Application) Intermittent Handling of Hot, Dry Objects
Glove Cuff Style Knit
Series 43-113
Yarn Gauge 7
Show More Show Less