missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercuric Nitrate, 0.141 N, 0.0705 M, 1 mg = 5 mL Cl-, Ricca Chemical
APHA/EPA for Chloride Testing
Supplier: Ricca Chemical Company 474016
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Specifications
| 500 mL | |
| MFCD00149736 | |
| Amber Glass Bottle | |
| mercury(2+) hydrate dinitrate | |
| 342.62 | |
| Mercuric Nitrate |
| H2HgN2O7 | |
| KVICROHOONHSRH-UHFFFAOYSA-N | |
| O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 500 mL | |
| APHA/EPA for Chloride |