missing translation for 'onlineSavingsMsg'
Learn More

(+)-Menthyl Chloroformate 97.0+%, TCI America™

Catalog No. M12215ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12215ML 5 mL
M122125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12215ML Supplier TCI America Supplier No. M12215ML
Only null left

Chemical Identifiers

CAS 7635-54-3
Molecular Formula C11H19ClO2
Molecular Weight (g/mol) 218.721
MDL Number MFCD00134483
InChI Key KIUPCUCGVCGPPA-IDKOKCKLSA-N
Synonym 1s-+-menthyl chloroformate
PubChem CID 57369944
IUPAC Name [(5S)-5-methyl-2-propan-2-ylcyclohexyl] carbonochloridate
SMILES CC1CCC(C(C1)OC(=O)Cl)C(C)C

Specifications

Color Colorless
Quantity 5 mL
UN Number 3277
Formula Weight 218.72
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material (+)-Menthyl Chloroformate