missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Menthoxyacetic Acid 98.0+%, TCI America™
Supplier: TCI America M05735G
Specifications
| (-)-Menthoxyacetic Acid | |
| 54°C | |
| C12H22O3 | |
| 5 g | |
| CC1CCC(C(C1)OCC(=O)O)C(C)C | |
| 214.305 | |
| 214.31 | |
| Crystalline Powder |
| 40248-63-3 | |
| Yellow | |
| MFCD00001483 | |
| CILPHQCEVYJUDN-MXWKQRLJSA-N | |
| 2-[(1S,2R,5R)-5-methyl-2-propan-2-ylcyclohexyl]oxyacetic acid | |
| 25277152 | |
| ≥98.0% (T) |
Chemical Identifiers
| 40248-63-3 | |
| 214.305 | |
| CILPHQCEVYJUDN-MXWKQRLJSA-N | |
| 2-[(1S,2R,5R)-5-methyl-2-propan-2-ylcyclohexyl]oxyacetic acid |
| C12H22O3 | |
| MFCD00001483 | |
| 25277152 | |
| CC1CCC(C(C1)OCC(=O)O)C(C)C |
Safety and Handling
TSCA : No