missing translation for 'onlineSavingsMsg'
Learn More

(-)-Menthoxyacetic Acid 98.0+%, TCI America™

Supplier:  TCI America M057325G

 View more versions of this product

Catalog No. M057325G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

(-)-Menthoxyacetic Acid
54°C
C12H22O3
25 g
CC1CCC(C(C1)OCC(=O)O)C(C)C
214.305
214.31
Crystalline Powder
40248-63-3
Yellow
MFCD00001483
CILPHQCEVYJUDN-MXWKQRLJSA-N
2-[(1S,2R,5R)-5-methyl-2-propan-2-ylcyclohexyl]oxyacetic acid
25277152
≥98.0% (T)

Chemical Identifiers

40248-63-3
214.305
CILPHQCEVYJUDN-MXWKQRLJSA-N
2-[(1S,2R,5R)-5-methyl-2-propan-2-ylcyclohexyl]oxyacetic acid
C12H22O3
MFCD00001483
25277152
CC1CCC(C(C1)OCC(=O)O)C(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions