missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Menthone, Terpene Analytical Standard, MilliporeSigma™ Supelco™
Supplier: Merck Emd Millipore 6367725ML
Chemical Identifiers
| 14073-97-3 | |
| 207°C to 210°C (literature) | |
| C10H18O | |
| n20/D 1.45 (literature); n20/D 1.451 | |
| (1 R,4 S)-p-Menthan-3-one; (2 S,5 R)-2-Isopropyl-5-methylcyclohexanone | |
| 154.25 | |
| ≥99% (Sum of Enantiomers, GC) | |
| Neat |
| 0.893 g/mL (at 20°C (literature)) | |
| 74°C | |
| C10H18O | |
| MFCD00001634 | |
| [α]20/D -28 ±2°, Neat | |
| 0.5 mmHg (20°C) | |
| Analytical Standard |
Specifications
| 14073-97-3 | |
| 207°C to 210°C (literature) | |
| C10H18O | |
| C10H18O | |
| 25 mL | |
| [α]20/D -28 ±2°, Neat | |
| 0.5 mmHg (20°C) | |
| Analytical Standard |
| 0.893 g/mL (at 20°C (literature)) | |
| 74°C | |
| n20/D 1.45 (literature); n20/D 1.451 | |
| MFCD00001634 | |
| (1 R,4 S)-p-Menthan-3-one; (2 S,5 R)-2-Isopropyl-5-methylcyclohexanone | |
| 154.25 | |
| ≥99% (Sum of Enantiomers, GC) | |
| Neat |
Safety and Handling
H315 - H317
ShelfLife : Limited shelf life, expiry date on the label
Recommended Storage : 2°C to 8°C