missing translation for 'onlineSavingsMsg'
Learn More

Menthone, mixture of isomers, 98%

Catalog No. AAA1766518
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1766518 50 g
AAA1766530 250 g
AAA176650B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1766518 Supplier Thermo Scientific Chemicals Supplier No. A1766518

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10458-14-7 | C10H18O | 154.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10458-14-7
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00062998
InChI Key NFLGAXVYCFJBMK-UHFFFAOYSA-N
Synonym isomenthone, p-menthan-3-one, 2-isopropyl-5-methylcyclohexanone, menthone, cyclohexanone, 5-methyl-2-1-methylethyl, 5-methyl-2-1-methylethyl cyclohexanone, p-menthanone, 2-isopropyl-5-methyl-cyclohexanone, p-menthan-3-one, dl, 5-methyl-2-isopropyl cyclohexanone
PubChem CID 6986
ChEBI CHEBI:36742
IUPAC Name 5-methyl-2-propan-2-ylcyclohexan-1-one
SMILES CC1CCC(C(=O)C1)C(C)C

Specifications

Density 0.894
Boiling Point ∼207°C
Flash Point 72°C (162°F)
Quantity 50 g
Formula Weight 154.25
Percent Purity 98%
Chemical Name or Material Menthone, mixture of isomers
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 233-944-9
RTECSNumber OS9541500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only