missing translation for 'onlineSavingsMsg'
Learn More

Menthone, mixture of isomers, 98%

CAS: 10458-14-7 | C10H18O | 154.253 g/mol

Supplier:  Thermo Scientific Chemicals A176650B

Quantity 1000 g

 View more versions of this product

Catalog No. AAA176650B

Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10458-14-7
154.253
NFLGAXVYCFJBMK-UHFFFAOYSA-N
6986
5-methyl-2-propan-2-ylcyclohexan-1-one
C10H18O
MFCD00062998
isomenthone, p-menthan-3-one, 2-isopropyl-5-methylcyclohexanone, menthone, cyclohexanone, 5-methyl-2-1-methylethyl, 5-methyl-2-1-methylethyl cyclohexanone, p-menthanone, 2-isopropyl-5-methyl-cyclohexanone, p-menthan-3-one, dl, 5-methyl-2-isopropyl cyclohexanone
CHEBI:36742
CC1CCC(C(=O)C1)C(C)C

Specifications

Menthone
10458-14-7
∼207°C
C10H18O
MFCD00062998
NFLGAXVYCFJBMK-UHFFFAOYSA-N
5-methyl-2-propan-2-ylcyclohexan-1-one
6986
154.25
mixture of isomers
0.894
72°C (162°F)
1000 g
isomenthone, p-menthan-3-one, 2-isopropyl-5-methylcyclohexanone, menthone, cyclohexanone, 5-methyl-2-1-methylethyl, 5-methyl-2-1-methylethyl cyclohexanone, p-menthanone, 2-isopropyl-5-methyl-cyclohexanone, p-menthan-3-one, dl, 5-methyl-2-isopropyl cyclohexanone
CC1CCC(C(=O)C1)C(C)C
154.253
CHEBI:36742
98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 233-944-9

RTECSNumber : OS9541500

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions