missing translation for 'onlineSavingsMsg'
Learn More

Menthofuran, 95%

CAS: 494-90-6 | C10H14O | 150.221 g/mol

$87.04 - $186.29

Chemical Identifiers

CAS 494-90-6
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00041851
InChI Key YGWKXXYGDYYFJU-UHFFFAOYSA-N
Synonym menthofuran, menthofurane, 3,9-epoxy-p-mentha-3,8-diene, benzofuran, 4,5,6,7-tetrahydro-3,6-dimethyl, p-mentha-3,8-diene, 3,9-epoxy, 4,5,6,7-tetrahydro-3,6-dimethylbenzofuran, fema no. 3235, 4,5,6,7-tetrahydro-3,6-dimethylcoumarone, r-menthofuran, 3,6-dimethyl-4,5,6,7-tetrahydrobenzo 2,1-b furan
PubChem CID 329983
ChEBI CHEBI:50542
IUPAC Name 3,6-dimethyl-4,5,6,7-tetrahydro-1-benzofuran
SMILES CC1CCC2=C(C1)OC=C2C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAA1719203
View Documents Promotion Details
N/A 1 g
Each for $87.04
Only null left
 
AAA1719206
View Documents Promotion Details
N/A 5 g
Each for $186.29
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

494-90-6
150.221
YGWKXXYGDYYFJU-UHFFFAOYSA-N
329983
3,6-dimethyl-4,5,6,7-tetrahydro-1-benzofuran
C10H14O
MFCD00041851
menthofuran, menthofurane, 3,9-epoxy-p-mentha-3,8-diene, benzofuran, 4,5,6,7-tetrahydro-3,6-dimethyl, p-mentha-3,8-diene, 3,9-epoxy, 4,5,6,7-tetrahydro-3,6-dimethylbenzofuran, fema no. 3235, 4,5,6,7-tetrahydro-3,6-dimethylcoumarone, r-menthofuran, 3,6-dimethyl-4,5,6,7-tetrahydrobenzo 2,1-b furan
CHEBI:50542
CC1CCC2=C(C1)OC=C2C

Specifications

494-90-6
86°C to 88°C (15 mmHg)
C10H14O
MFCD00041851
YGWKXXYGDYYFJU-UHFFFAOYSA-N
3,6-dimethyl-4,5,6,7-tetrahydro-1-benzofuran
329983
150.22
Menthofuran
0.972
75°C (167°F)
1.4845
menthofuran, menthofurane, 3,9-epoxy-p-mentha-3,8-diene, benzofuran, 4,5,6,7-tetrahydro-3,6-dimethyl, p-mentha-3,8-diene, 3,9-epoxy, 4,5,6,7-tetrahydro-3,6-dimethylbenzofuran, fema no. 3235, 4,5,6,7-tetrahydro-3,6-dimethylcoumarone, r-menthofuran, 3,6-dimethyl-4,5,6,7-tetrahydrobenzo 2,1-b furan
CC1CCC2=C(C1)OC=C2C
150.221
CHEBI:50542
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c

H227-H315-H319

EINECSNumber : 207-795-5

RTECSNumber : DF7995750

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only