missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Men's Charcoal Lightweight Long Sleeve Flame Resistant T-shirt
Description
- Underarm gusset for improved breathability
- Single spade chest pocket
- Rib-knit crewneck collar
- Hazard: Flash Fire, Arc Flash
- Wash Care: Home Wash
- Fabric: EXCEL FR™ Flame resistant, 7 oz. (235 g/m2) Interlock
- Blend: 100% Cotton
- Country of Origin: Imported, Made in USA Fabric
Specifications
Specifications
| Size | Medium |
| Gender | Men's |
| Material | FR Cotton |
| Color | Charcoal |
| Neck Style | Hemmed Crew |
| Sleeve Style | Long Sleeve |
| Closure Type | Pull On |
| Disposable | Reusable |
| No. of Pockets | 1 |
| Hazard Risk Category | Category 2 |
| Show More |