missing translation for 'onlineSavingsMsg'
Learn More
Learn More
- Home
- Products
- Personal Protective Equipment
- Safety Clothing
- Pants, Trousers, and Leggings
- Heat and Fire Resistant Pants, Trousers, and Leggings
- Bulwark™ Men's Stretch Denim Flame Resistant Dungaree
Bulwark™ Men's Stretch Denim Flame Resistant Dungaree
Catalog No. 17204233
Change view
Click to view available options
Size:
30 x 30 in.
30 x 32 in.
30 x 34 in.
32 x 30 in.
32 x 32 in.
32 x 34 in.
32 x 36 in.
33 x 30 in.
33 x 32 in.
33 x 34 in.
33 x 36 in.
34 x 30 in.
34 x 32 in.
34 x 34 in.
34 x 36 in.
35 x 30 in.
35 x 32 in.
35 x 34 in.
35 x 36 in.
36 x 32 in.
36 x 34 in.
36 x 36 in.
38 x 30 in.
38 x 32 in.
38 x 34 in.
38 x 36 in.
40 x 30 in.
40 x 32 in.
40 x 34 in.
40 x 36 in.
42 x 30 in.
42 x 32 in.
42 x 34 in.
42 x 36 in.
44 x 30 in.
44 x 32 in.
44 x 34 in.
44 x 36 in.
46 x 30 in.
46 x 32 in.
46 x 34 in.
48 x 30 in.
48 x 32 in.
50 x 30 in.
50 x 32 in.
50 x 34 in.
46 product options available for selection
| Catalog No. | Size |
|---|---|
| 17204233 | 30 x 30 in. |
| 17203943 | 30 x 32 in. |
| 17204032 | 30 x 34 in. |
| 17204222 | 32 x 30 in. |
| 17204224 | 32 x 32 in. |
| 17204050 | 32 x 34 in. |
| 17204165 | 32 x 36 in. |
| 17204363 | 33 x 30 in. |
| 17204453 | 33 x 32 in. |
| 17204371 | 33 x 34 in. |
| 17204372 | 33 x 36 in. |
| 17204230 | 34 x 30 in. |
| 17204020 | 34 x 32 in. |
| 17204215 | 34 x 34 in. |
| 17204121 | 34 x 36 in. |
| 17204446 | 35 x 30 in. |
| 17204249 | 35 x 32 in. |
| 17204448 | 35 x 34 in. |
| 17204132 | 35 x 36 in. |
| 17204350 | 36 x 32 in. |
| 17204258 | 36 x 34 in. |
| 17204266 | 36 x 36 in. |
| 17204314 | 38 x 30 in. |
| 17203972 | 38 x 32 in. |
| 17204023 | 38 x 34 in. |
| 17204478 | 38 x 36 in. |
| 17203961 | 40 x 30 in. |
| 17203963 | 40 x 32 in. |
| 17203978 | 40 x 34 in. |
| 17204316 | 40 x 36 in. |
| 17203985 | 42 x 30 in. |
| 17204475 | 42 x 32 in. |
| 17204257 | 42 x 34 in. |
| 17203974 | 42 x 36 in. |
| 17204275 | 44 x 30 in. |
| 17204309 | 44 x 32 in. |
| 17204262 | 44 x 34 in. |
| 17203964 | 44 x 36 in. |
| 17204174 | 46 x 30 in. |
| 17204279 | 46 x 32 in. |
| 17204473 | 46 x 34 in. |
| 17203988 | 48 x 30 in. |
| 17204271 | 48 x 32 in. |
| 17204307 | 50 x 30 in. |
| 17204467 | 50 x 32 in. |
| 17103758 | 50 x 34 in. |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 46 options available.
Description
- Increased durability with triple needle felled out seams and seat seam
- Sits below natural waist with relaxed fit through waist, hip, and thigh
- Bootcut leg opening fits easily over boots
- Five traditional jean style pockets, with an additional ruler pocket on right leg and utility pocket on left leg for extra storage
- One-piece waistband with metal button closure (metal is not exposed to skin)
- Utility hammer loop on left hip
- Reinforced pockets help prevent holes
- Hazard: Arc Flash, Flash Fire
- Number of Pockets: 7
- Primary Closure: Concealed Button
- Wash Care: Industrial Laundry - Light Soil
- Fabric: Body: 12 oz. (405 g/m2) Flame Resistant Denim
- Blend: Body: 94% Cotton/5% Polyester/1% Spandex Elastane
- Country of Origin: Imported, Made in USA Fabric