missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Men's Navy Flame Resistant Flex Knit Long Sleeve T-shirt

Catalog No. 17204306
Change view
Click to view available options
Size:
2X-Large (Long)
Large (Long)
X-Large (Long)
Small
Medium
Large
X-Large
2X-Large
3X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17204306 Small
17204479 Medium
17204415 Large
17204466 X-Large
17204312 2X-Large
17204471 3X-Large
17204264 Large (Long)
17204172 X-Large (Long)
17103740 2X-Large (Long)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 17204306 Supplier Bulwark™ Supplier No. STT2NV RG S

Men's Navy Flame Resistant Flex Knit Long Sleeve T-shirt gives you the stretch of your favorite T-shirt while keeping you cool and dry.

  • Lightweight fabric flexes for added comfort and range of movement
  • Contrasting pattern on sleeve for an athletic look
  • Curved flatlock seams for optimal fit
  • Unique front pocket design with pencil stall
  • Ribbed collar
  • CAT 2/2112 left sleeve label leaves no question that you are protected on the job site
  • Hazard: Flash Fire, Arc Flash
  • Wash Care: Home Wash
  • Fabric: Body: 6.75 oz. (230 g/m&superscript_2;) Flame Resistant Jersey, Contrast Panels: 6.75 oz. (230 g/m2) Flame Resistant Jersey
  • Blend: Body: 93% Cotton/7% Spandex Elastane, Contrast Panels: 93% Cotton/7% Spandex Elastane
  • Country of Origin: Imported, Made in USA Fabric

Specifications

Size Small
Gender Men's
Material Cotton, Spandex
Color Navy
Neck Style Hemmed Crew
Sleeve Style Long Sleeve
Construction Knit
Closure Type Button
Disposable Reusable
No. of Pockets 1
Hazard Risk Category Category 2
Arc Flash Rating 8.3
Certifications/Compliance NFPA 2112, NFPA 70E
Show More Show Less