missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Men's Navy Flame Resistant Flex Knit Long Sleeve T-shirt
Men's Navy Flame Resistant Flex Knit Long Sleeve T-shirt gives you the stretch of your favorite T-shirt while keeping you cool and dry.
Supplier: Bulwark™ STT2NVLNXL
Description
- Lightweight fabric flexes for added comfort and range of movement
- Contrasting pattern on sleeve for an athletic look
- Curved flatlock seams for optimal fit
- Unique front pocket design with pencil stall
- Ribbed collar
- CAT 2/2112 left sleeve label leaves no question that you are protected on the job site
- Hazard: Flash Fire, Arc Flash
- Wash Care: Home Wash
- Fabric: Body: 6.75 oz. (230 g/m&superscript_2;) Flame Resistant Jersey, Contrast Panels: 6.75 oz. (230 g/m2) Flame Resistant Jersey
- Blend: Body: 93% Cotton/7% Spandex Elastane, Contrast Panels: 93% Cotton/7% Spandex Elastane
- Country of Origin: Imported, Made in USA Fabric
Specifications
| X-Large (Long) | |
| Cotton, Spandex | |
| Hemmed Crew | |
| Knit | |
| Reusable | |
| Category 2 | |
| NFPA 2112, NFPA 70E |
| Men's | |
| Navy | |
| Long Sleeve | |
| Button | |
| 1 | |
| 8.3 |