missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Men's Navy Flame Resistant Flex Knit Button Down Shirt

Catalog No. 17204412
Change view
Click to view available options
Size:
2X-Large (Long)
Large (Long)
Medium (Long)
X-Large (Long)
Small
Medium
Large
X-Large
2X-Large
3X-Large
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17204412 Medium
17204025 Small
17203971 Large
17203960 X-Large
17204401 2X-Large
17204239 3X-Large
17103748 Medium (Long)
17204084 Large (Long)
17204089 X-Large (Long)
17204182 2X-Large (Long)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 17204412 Supplier Bulwark™ Supplier No. STG4NV RG M

Men's Navy Flame Resistant Flex Knit Button Down Shirt combines the crisp, professional look of an office work shirt and the comfort of your favorite T-shirt.

  • Spandex-blend fabric and side seam gussets allows it to stretch freely without coming untucked
  • Two reinforced chest pockets with button flaps allows convenient storage
  • Banded collar with full underarm gusset for added mobility
  • Two chest pockets with flaps and button closure
  • Lightweight, stretch fabric
  • Easy-to-spot CAT 2/2112 label on the left sleeve leave no question that you're protected on the job site
  • Hazard: Arc Flash, Flash Fire
  • Wash Care: Industrial Laundry - Light Soil
  • Fabric: Body: 6.75 oz. (230 g/m2) Flame Resistant Jersey
  • Blend: Body: 93% Cotton/7% Spandex Elastane
  • Country of Origin: Imported, Made in USA Fabric

Specifications

Size Medium
Gender Men's
Material Cotton, Spandex
Color Navy
Neck Style Collar
Sleeve Style Long Sleeve
Construction Knit
Closure Type Button
Disposable Reusable
No. of Pockets 2
Hazard Risk Category Category 2
Arc Flash Rating 8.3
Certifications/Compliance ASTM F1506, NFPA 70E, NFPA 2112
Show More Show Less