missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Men's Midweight Excel FR™ ComforTouch™ Navy Premium Flame Resistant Coverall

Catalog No. 17204184
Change view
Click to view available options
Size:
2X-Large (Long)
3X-Large (Long)
4X-Large (Long)
Large (Long)
X-Large (Long)
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17204184 Medium
17204244 Large
17204017 X-Large
17204099 2X-Large
17204393 3X-Large
17204183 4X-Large
17204366 Large (Long)
17204136 X-Large (Long)
17204139 2X-Large (Long)
17204374 3X-Large (Long)
17204124 4X-Large (Long)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 17204184 Supplier Bulwark™ Supplier No. CLB6NV RG M

Men's Midweight Excel FR™ ComforTouch™ Navy Premium Flame Resistant Coverall is a midweight premium coverall provides CAT 2 FR/AR protection with innovative features for extra comfort and convenience.

  • This midweight premium coverall provides CAT 2 FR/AR protection with innovative features for extra comfort and convenience.
  • Eight total pockets keep your tools at hand, while elastic waist inserts and deep-pleated action back provide added mobility and comfort while you work.
  • Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Flash Fire, Arc Flash
  • Primary Closure: Concealed Zipper
  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: Flame resistant, 9 oz. (305 g/m&superscript_2;) Twill
  • Blend: 88% Cotton/ 12% Nylon
  • Closure: Two-way concealed Nomex™ taped brass break-away zipper, concealed snap at top of zipper and at neck
  • Pocket: Two front swing pockets are topstitched and lined and two patch hip pockets have single concealed snap closure, One left sleeve pocket, two patch chest pockets have flaps and concealed snap closure and a rule pocket on right leg
  • Features: Two-way concealed Nomex™ taped brass break-away zipper, concealed snap at top of zipper and at neck, Two-piece, lay-flay collar, deep-pleated action back and elastic waist inserts, Hemmed sleeve with adjustable snap and side vent opening with concealed snap closure
  • Product Collection: Excel FR Comfortouch
  • Protection: ATPV Arc Rating: 12
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Designed to be worn over other clothing. Fit over work boots.
  • Gender: Male
  • Materials: Cotton/Nylon
  • Brand: Bulwark™

Specifications

Product Type Flame-resistant Coveralls
Size Medium
Gender Men's
Material Cotton, Nylon
Color Navy
Disposable Reusable
Waist Style Elastic (Stitched-in)
Ankle Style Open
Cuff Style Open
Neck Style Collared
Hood Type Unhooded
Closure Type 2-way Front Zipper with Storm Flap
Seam Type Stitched
Sterility Non-sterile
Protection Class CAT 2
Resistance Flame
Arc Flash Rating 12
Certifications/Compliance NFPA 2112
No. of Pockets 8
Show More Show Less