missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Men's Midweight Excel FR™ ComforTouch™ Deluxe Insulated Bib Overall
Men's Midweight Excel FR™ ComforTouch™ Deluxe Insulated Bib Overall offers unbeatable warmth with CAT 2 AR/FR protection and deluxe features like rugged, double-reinforced knees. Hook and loop leg tabs ensure your coverall fits snugly around your boots.
Supplier: Bulwark™ BLC8GYLNL
Description
- For cold jobs, this midweight insulated bib overall offers unbeatable warmth with CAT 3 AR/FR protection and deluxe features like rugged, double-reinforced knees.
- Large patch pockets and bonus utility pockets ensure there's a place for all of your tools.
- Certifications Level 1: NFPA 2112, NFPA 70E
- Certifications Level 2: Category 3 Protection, NFPA 2112 - 2018 Compliant
- Hazard: Flash Fire, Arc Flash
- Primary Closure: Concealed Zipper
- Wash Care: Industrial Laundry - Light Soil, Home Wash
Fabric: Shell: Flame resistant, 7 oz. (235 g/m&superscript_2;) Twill, Water-repellent, Insulation Package: 14 oz. (474 g/m&superscript_2;) - Blend: Shell: 88% Cotton/ 12% Nylon, Insulation Package: 50% Modacrylic 50% Cotton
- Closure: Concealed Nomex™ taped brass zipper front
- Pocket: Two large front bib pockets with pencil stall, Two large hip pockets, One rule pocket on right leg, Two front pockets
- Features: Reinforced double knees, Elasticized shoulder straps with non-conductive adjustable hardware, Two large center front pockets with pencil stall
- Product Collection: Excel FR Comfortouch
- Protection: ATPV Arc Rating: 36
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Designed to be worn over other clothing. Fit over work boots.
- Gender: Male
- Materials: Cotton/Nylon
- Brand: Bulwark™
Specifications
| Zipper | |
| Cotton, Nylon | |
| Large (Long) | |
| 36 |
| Gray | |
| Stitched | |
| Adjustable | |
| NFPA 2112, NFPA 70E |