missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Men's Long Sleeve Navy Flame Resistant Performance T-Shirt

Catalog No. 17204263
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17204263 2X-Large
17204255 Medium
17204024 Large
17203959 X-Large
17203984 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17204263 Supplier Bulwark™ Supplier No. SMT2NV RG XXL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Men's Long Sleeve Navy Flame Resistant Performance T-Shirt provides flame-resistant protection in a lightweight blended fabric. The result is unbeatable softness coupled with lasting durability. This casual FR shirt features raglan sleeves as well as rib knit collar and cuffs

  • As part of our CoolTouch™ 2 fabric collection, this long sleeve tee provides flame-resistant protection in a lightweight blended fabric.
  • The result is unbeatable softness coupled with lasting durability.
  • This casual FR shirt features raglan sleeves as well as rib knit collar and cuffs
  • Certifications Level 1: NFPA 2112, NFPA 70E
  • Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Flash Fire, Arc Flash
  • Number of Pockets: 1
  • Primary Closure: No Closure
  • Wash Care: Industrial Laundry - Light Soil, Home Wash
  • Fabric: Flame-resistant, 6.5 oz. (220 g/m&superscript_2;) with moisture management finish
  • Blend: 45% Modacrylic / 35% FR Lyocell / 20% Aramid
  • Collar: Rib knit collar
  • Pocket: Single spade chest pocket
  • Features: Heat seal Bulwark™ triangle centered at neck to show wearer has on FR undergarment, Full side seam gusset, Rib knit cuffs and collar, Raglan sleeves for ease of movement, Tagless brand label and warning label
  • Product Collection: Cooltouch 2
  • Packaging: Hang tag
  • Protection: EBT Arc Rating: 8.2
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Regular Fit
  • Gender: Male
  • Materials: Modacrylic/FR Lyocell/Aramid
  • Brand: Bulwark™

Specifications

Size 2X-Large
Gender Men's
Material Modacrylic Blend
Color Navy
Neck Style Hemmed Crew
Sleeve Style Raglan Sleeve
Closure Type Pull On
Disposable Reusable
No. of Pockets 1
Hazard Risk Category Category 2
Arc Flash Rating 8.2
Certifications/Compliance NFPA 2112, NFPA 70E
Show More Show Less