missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Men's Long Sleeve Navy Flame Resistant Performance T-Shirt
Men's Long Sleeve Navy Flame Resistant Performance T-Shirt provides flame-resistant protection in a lightweight blended fabric. The result is unbeatable softness coupled with lasting durability. This casual FR shirt features raglan sleeves as well as rib knit collar and cuffs
Supplier: Bulwark™ SMT2NVRG3XL
Description
- As part of our CoolTouch™ 2 fabric collection, this long sleeve tee provides flame-resistant protection in a lightweight blended fabric.
- The result is unbeatable softness coupled with lasting durability.
- This casual FR shirt features raglan sleeves as well as rib knit collar and cuffs
- Certifications Level 1: NFPA 2112, NFPA 70E
- Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
- Hazard: Flash Fire, Arc Flash
- Number of Pockets: 1
- Primary Closure: No Closure
- Wash Care: Industrial Laundry - Light Soil, Home Wash
- Fabric: Flame-resistant, 6.5 oz. (220 g/m&superscript_2;) with moisture management finish
- Blend: 45% Modacrylic / 35% FR Lyocell / 20% Aramid
- Collar: Rib knit collar
- Pocket: Single spade chest pocket
- Features: Heat seal Bulwark™ triangle centered at neck to show wearer has on FR undergarment, Full side seam gusset, Rib knit cuffs and collar, Raglan sleeves for ease of movement, Tagless brand label and warning label
- Product Collection: Cooltouch 2
- Packaging: Hang tag
- Protection: EBT Arc Rating: 8.2
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Regular Fit
- Gender: Male
- Materials: Modacrylic/FR Lyocell/Aramid
- Brand: Bulwark™
Specifications
| 3X-Large | |
| Modacrylic Blend | |
| Hemmed Crew | |
| Pull On | |
| 1 | |
| 8.2 |
| Men's | |
| Navy | |
| Raglan Sleeve | |
| Reusable | |
| Category 2 | |
| NFPA 2112, NFPA 70E |