missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Men's Lightweight Navy Flame Resistant T-Shirt
Men's Lightweight Navy Flame Resistant T-Shirt is a CAT 2 AR/FR, NFPA™ 2112 compliant tee that features a heat seal Bulwark™ triangle centered at neck to show wearer has on FR undergarment.
Supplier: Bulwark™ SET8NVSSXXL
Description
- Meet the most comfortable FR short sleeve T-shirt you'll ever wear.
- This CAT 2 AR/FR, NFPA™ 2112 compliant tee features a heat seal Bulwark™ triangle centered at neck to show wearer has on FR undergarment.
- Certifications Level 1: ASTM F1506, NFPA 2112, NFPA 70E
- Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
- Hazard: Flash Fire, Arc Flash
- Primary Closure: No Closure
- Wash Care: Home Wash
- Fabric: EXCEL FR™ Flame resistant, 7 oz. (235 g/m&superscript_2;) Interlock
- Blend: 100% Cotton
- Collar: Rib-knit crewneck collar
- Pocket: Single spade chest pocket
- Features: Hemmed sleeve, Heat seal Bulwark™ triangle centered at neck to show wearer has FR undergarment, Underarm gusset for improved breathability
- Product Collection: Excel FR
- Protection: ATPV Arc Rating: 8.9
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Regular Fit
- Gender: Male
- Materials: Cotton
- Brand: Bulwark™
Specifications
| 2X-Large | |
| FR Cotton | |
| Hemmed Crew | |
| Pull On | |
| 1 | |
| 8.9 |
| Men's | |
| Navy | |
| Short Sleeve | |
| Reusable | |
| Category 2 | |
| ASTM F1506, NFPA 2112, NFPA 70E |