missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Men's Lightweight Khaki Flame Resistant Cargo Pant

Catalog No. 17204118
Change view
Click to view available options
Size:
31 x 37 in.
33 x 37 in.
34 x 37 in.
35 x 37 in.
36 x 37 in.
38 x 37 in.
40 x 37 in.
42 x 37 in.
50 x 37 in.
52 x 37 in.
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17204118 31 x 37 in.
17204440 33 x 37 in.
17204000 34 x 37 in.
17204178 35 x 37 in.
17204200 36 x 37 in.
17203981 38 x 37 in.
17103762 40 x 37 in.
17204253 42 x 37 in.
17204166 50 x 37 in.
17204259 52 x 37 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 17204118 Supplier Bulwark™ Supplier No. PMU2KH 31 37U

Men's Lightweight Khaki Flame Resistant Cargo Pant has six total pockets, including a cargo pocket on each leg to ensure you have plenty of room to store all your tools.

  • Ideal for storage and breathability
  • Sits at the waist
  • Regular fit in thigh and seat with slightly tapered leg
  • Two front slack-style pockets and two cargo utility pockets
  • Left cargo pocket has two snap closures and right utility pocket is open
  • Hazard: Flash Fire, Arc Flash
  • Waistband with hook and eye closure
  • Wash Care: Industrial Laundry - Light Soil, Home Wash
  • Fabric: Flame resistant, 7 oz. (235 g/m2)
  • Blend: 48% Modacrylic/37% Lyocell/15% Aramid
  • Product Collection: Cooltouch 2
  • Country of Origin: Imported, Made in USA Fabric

Specifications

Size 31 x 37 in.
Gender Men's
Material Modacrylic, Lyocell, Aramid
Color Khaki
Seam Type Stitched
No. of Pockets 6
Closure Type Hook and Eye
Disposable Reusable
Hazard Risk Category Category 2
Arc Flash Rating 9
Certifications/Compliance NFPA 2112
Show More Show Less