missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Men's Lightweight Khaki Flame Resistant Cargo Pant
Men's Lightweight Khaki Flame Resistant Cargo Pant has six total pockets, including a cargo pocket on each leg to ensure you have plenty of room to store all your tools.
Supplier: Bulwark™ PMU2KH3837U
Description
- Ideal for storage and breathability
- Sits at the waist
- Regular fit in thigh and seat with slightly tapered leg
- Two front slack-style pockets and two cargo utility pockets
- Left cargo pocket has two snap closures and right utility pocket is open
- Hazard: Flash Fire, Arc Flash
- Waistband with hook and eye closure
- Wash Care: Industrial Laundry - Light Soil, Home Wash
- Fabric: Flame resistant, 7 oz. (235 g/m2)
- Blend: 48% Modacrylic/37% Lyocell/15% Aramid
- Product Collection: Cooltouch 2
- Country of Origin: Imported, Made in USA Fabric
Specifications
| 38 x 37 in. | |
| Modacrylic, Lyocell, Aramid | |
| Stitched | |
| Reusable | |
| 9 |
| Men's | |
| Khaki | |
| Hook and Eye | |
| Category 2 | |
| NFPA 2112 |