missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Men's Lightweight Excel FR™ ComfortTouch™ Khaki Premium Flame Resistant Coverall
Description
- Designed to be worn over clothing
- Two front swing pockets are topstitched and lined and two patch hip pockets have single concealed snap closure
- One left sleeve pocket, two patch chest pockets have flaps and concealed snap closure and a rule pocket on right leg
- Two-way concealed Nomex™ taped brass break-away zipper, concealed snap at top of zipper and at neck
- Hemmed sleeve with adjustable snap and side vent opening with concealed snap closure
- Fits over work boots
- Hazard: Flash Fire, Arc Flash
- Primary Closure: Concealed Zipper
- Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
- Fabric: Flame resistant, 7 oz. (235 g/m2)
- Blend: 88% Cotton/12% Nylon
- Country of Origin: Imported, Made in USA Fabric
Specifications
Specifications
| Product Type | Flame-resistant Coveralls |
| Size | Large |
| Gender | Men's |
| Material | Cotton, Nylon |
| Color | Khaki |
| Disposable | Reusable |
| Waist Style | Elastic (Stitched-in) |
| Ankle Style | Open |
| Cuff Style | Open |
| Neck Style | Collared |
| Show More |