missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Men's Lightweight Charcoal Flame Resistant T-Shirt

Catalog No. p-200000043
Change view
Click to view available options
Size:
2X-Large (Long)
Large (Long)
X-Large (Long)
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17204133 Small
17203950 Medium
17203949 Large
17204066 X-Large
17204456 2X-Large
17204218 3X-Large
17204288 4X-Large
17204134 Large (Long)
17204043 X-Large (Long)
17204291 2X-Large (Long)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 17204133 Supplier Bulwark™ Supplier No. SET8CH SS S
Only null left

Men's Lightweight Charcoal Flame Resistant T-Shirt is a CAT 2 AR/FR, NFPA™ 2112 compliant tee that features a heat seal Bulwark™ triangle centered at neck to show wearer has on FR undergarment.

  • Meet the most comfortable FR short sleeve T-shirt you'll ever wear.
  • This CAT 2 AR/FR, NFPA™ 2112 compliant tee features a heat seal Bulwark™ triangle centered at neck to show wearer has on FR undergarment.
  • Certifications Level 1: ASTM F1506, NFPA 2112, NFPA 70E
  • Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Flash Fire, Arc Flash
  • Primary Closure: No Closure
  • Wash Care: Home Wash
  • Fabric: EXCEL FR™ Flame resistant, 7 oz. (235 g/m&superscript_2;) Interlock
  • Blend: 100% Cotton
  • Collar: Rib-knit crewneck collar
  • Pocket: Single spade chest pocket
  • Features: Hemmed sleeve, Heat seal Bulwark™ triangle centered at neck to show wearer has FR undergarment, Underarm gusset for improved breathability
  • Product Collection: Excel FR
  • Protection: ATPV Arc Rating: 8.9
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Regular Fit
  • Gender: Male
  • Materials: Cotton
  • Brand: Bulwark™

Specifications

Size Small
Gender Men's
Material FR Cotton
Color Charcoal
Neck Style Hemmed Crew
Sleeve Style Short Sleeve
Closure Type Pull On
Disposable Reusable
No. of Pockets 1
Hazard Risk Category Category 2
Arc Flash Rating 8.9
Certifications/Compliance ASTM F1506, NFPA 2112, NFPA 70E
Show More Show Less