missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Men's Khaki Flame Resistant Flex Knit Long Sleeve T-Shirt

Men's Khaki Flame Resistant Flex Knit Long Sleeve T-Shirt gives you the stretch of your favorite T-shirt, while keeping you cool and dry.

Supplier:  Bulwark™ STT2KHRGXXL

 View more versions of this product

Catalog No. 17-204-303

Only null left
Explore available promotions

Description

Description

  • Bulwark FR Men's Flex Knit tee gives you the stretch of your favorite T-shirt while keeping you cool and dry.
  • The lightweight fabric flexes for added comfort and range of movement, curved flatlock seams for optimal fit, unique front pocket design with pencil stall.
  • CAT 2/2112 left sleeve label leaves no question you're protected on the job site.
  • Certifications Level 1: NFPA 2112, NFPA 70E
  • Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Flash Fire, Arc Flash
  • Number of Pockets: 1
  • Primary Closure: Pull On
  • Wash Care: Home Wash
  • Fabric: Body: 6.75 oz. (230 g/m&superscript_2;) Flame Resistant Jersey, Contrast Panels: 6.75 oz. (230 g/m&superscript_2;) Flame Resistant Jersey
  • Blend: Body: 93% Cotton / 7% Spandex Elastane, Contrast Panels: 93% Cotton / 7% Spandex Elastane
  • Collar: Rib
  • Pocket: Single chest pocket
  • Features: Contrasting pattern on sleeve for an athletic look, Lightweight stretch fabric flexes with your body for added comfort and range of movement
  • Protection: ATPV Arc Rating: 8.3
  • Country of Origin: Imported, Made in USA Fabric
  • Gender: Male
  • Brand: Bulwark™
Specifications

Specifications

2X-Large
Cotton, Spandex
Hemmed Crew
Knit
Reusable
Category 2
NFPA 2112, NFPA 70E
Men's
Khaki
Long Sleeve
Pull On
1
8.3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions