missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Men's Khaki Flame Resistant Flex Knit Long Sleeve T-Shirt
Men's Khaki Flame Resistant Flex Knit Long Sleeve T-Shirt gives you the stretch of your favorite T-shirt, while keeping you cool and dry.
Supplier: Bulwark™ STT2KHLNXL
Description
- Bulwark FR Men's Flex Knit tee gives you the stretch of your favorite T-shirt while keeping you cool and dry.
- The lightweight fabric flexes for added comfort and range of movement, curved flatlock seams for optimal fit, unique front pocket design with pencil stall.
- CAT 2/2112 left sleeve label leaves no question you're protected on the job site.
- Certifications Level 1: NFPA 2112, NFPA 70E
- Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
- Hazard: Flash Fire, Arc Flash
- Number of Pockets: 1
- Primary Closure: Pull On
- Wash Care: Home Wash
- Fabric: Body: 6.75 oz. (230 g/m&superscript_2;) Flame Resistant Jersey, Contrast Panels: 6.75 oz. (230 g/m&superscript_2;) Flame Resistant Jersey
- Blend: Body: 93% Cotton / 7% Spandex Elastane, Contrast Panels: 93% Cotton / 7% Spandex Elastane
- Collar: Rib
- Pocket: Single chest pocket
- Features: Contrasting pattern on sleeve for an athletic look, Lightweight stretch fabric flexes with your body for added comfort and range of movement
- Protection: ATPV Arc Rating: 8.3
- Country of Origin: Imported, Made in USA Fabric
- Gender: Male
- Brand: Bulwark™
Specifications
| X-Large (Long) | |
| Cotton, Spandex | |
| Hemmed Crew | |
| Knit | |
| Reusable | |
| Category 2 | |
| NFPA 2112, NFPA 70E |
| Men's | |
| Khaki | |
| Long Sleeve | |
| Pull On | |
| 1 | |
| 8.3 |