missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Men's Khaki Flame Resistant Flex Knit Henley

Catalog No. 17204282
Change view
Click to view available options
Size:
2X-Large (Long)
Large (Long)
X-Large (Long)
Small
Medium
Large
X-Large
2X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17204282 Small
17204436 Medium
17103743 Large
17204011 X-Large
17204325 2X-Large
17103745 Large (Long)
17204234 X-Large (Long)
17103744 2X-Large (Long)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17204282 Supplier Bulwark™ Supplier No. STL2KH RG S

Men's Khaki Flame Resistant Flex Knit Henley has a casual, contemporary look. The lightweight fabric flexes for added comfort and range of movement, while curved flatlock seams allow for an optimal fit.

  • Unique front pocket design with pencil stall
  • Three-button front placket for enhanced appearance
  • Contrasting pattern on sleeve for athletic look
  • CAT 2/2112 left sleeve label leaves no question you're protected on the job site
  • Hazard: Flash Fire, Arc Flash
  • Wash Care: Home Wash
  • Fabric: Body: 6.75 oz. (230 g/m2) Flame Resistant Jersey, Contrast Panels: 6.75 oz. (230 g/m2) Flame Resistant Jersey
  • Blend: Body: 93% Cotton/7% Spandex Elastane, Contrast Panels: 93% Cotton/7% Spandex Elastane
  • Collar: Rib
  • Country of Origin: Imported, Made in USA Fabric

Specifications

Size Small
Gender Men's
Material Cotton, Spandex
Color Khaki
Neck Style Henley
Sleeve Style Long Sleeve
Construction Knit
Closure Type Button
Disposable Reusable
No. of Pockets 1
Hazard Risk Category Category 2
Arc Flash Rating 8.3
Certifications/Compliance NFPA 2112, NFPA 70E
Show More Show Less