missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Men's Excel FR™ ComforTouch™ Grey Premium Flame Resistant Coverall
Men's Excel FR™ ComforTouch™ Grey Premium Flame Resistant Coverall provides CAT 2 FR/AR protection and extra warmth for cold weather environments, without added bulk.
Supplier: Bulwark™ CLC8GYRGL
Description
- This premium coverall provides CAT 3 FR/AR protection and extra warmth for cold weather environments, without added bulk.
- It also accommodates snap on FR hoods for additional insulation from the elements.
- Certifications Level 1: ASTM F1506, NFPA 2112, NFPA 70E
- Certifications Level 2: Category 3 Protection
- Hazard: Flash Fire, Arc Flash
- Primary Closure: Concealed Zipper
- Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
- Fabric: Shell: Flame resistant, 7 oz. (235 g/m&superscript_2;) Twill, Water-repellent, Insulation Package: 14 oz. (474 g/m&superscript_2;)
- Blend: Shell: 88% Cotton/ 12% Nylon, Lining: 100% Cotton, Insulation Package: 100% Modacrylic
- Closure: Two-way, extra heavy-duty Nomex™ taped brass zipper front with concealed snap closures
- Collar: Two-piece, insulated, stand-up collar with concealed snap closure
- Pocket: Seven large patch pockets, two with hook-and-loop closure
- Features: Full Nomex™ taped ankle-to-hip brass zippers, concealed snap closure
- Product Collection: Excel FR Comfortouch
- Protection: ATPV Arc Rating: 36
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Designed to be worn over other clothing. Fit over work boots.
- Gender: Male
- Materials: Cotton, Nylon
- Brand: Bulwark™
Specifications
| Flame-resistant Coveralls | |
| Men's | |
| Gray | |
| Elastic (Stitched-in) | |
| Open | |
| Unhooded | |
| Stitched | |
| CAT 3 | |
| Two-Piece, Insulated, Stand-Up Collar | |
| ASTM F1506, NFPA 2112, NFPA 70E |
| Large | |
| Cotton, Nylon | |
| Reusable | |
| Open | |
| Collared | |
| Snap | |
| Non-sterile | |
| Flame | |
| 36 | |
| 7 |