missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Men's Charcoal Plaid Flame Resistant Flex Knit Button Down Shirt

Men's Charcoal Plaid Flame Resistant Flex Knit Button Down Shirt provides flame-resistant protection in a lightweight blended fabric. The result is unbeatable softness coupled with lasting durability. This casual FR shirt features raglan sleeves as well as rib knit collar and cuffs.

$152.92 - $152.92

Specifications

Gender Men's
Material Cotton, Spandex
Color Charcoal
Neck Style Collar
Sleeve Style Long Sleeve
View More Specs

Products 10
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17-204-054
View Documents
Bulwark™
STG2CHRGS
Small
Each for $152.92
Only null left
 
17-204-096
View Documents
Bulwark™
STG2CHRGM
Medium
Each for $152.92
Only null left
 
17-204-091
View Documents
Bulwark™
STG2CHRGL
Large
Each for $152.92
Only null left
 
17-204-243
View Documents
Bulwark™
STG2CHRGXL
X-Large
Each for $152.92
Only null left
 
17-204-044
View Documents
Bulwark™
STG2CHRGXXL
2X-Large
Each for $152.92
Only null left
 
17-204-138
View Documents
Bulwark™
STG2CHLNM
Medium (Long)
Each for $152.92
Only null left
 
17-204-445
View Documents
Bulwark™
STG2CHLNL
Large (Long)
Each for $152.92
Only null left
 
17-204-225
View Documents
Bulwark™
STG2CHLNXL
X-Large (Long)
Each for $152.92
Only null left
 
17-203-957
View Documents
Bulwark™
STG2CHLNXXL
2X-Large (Long)
Each for $152.92
Only null left
 
17-204-295
View Documents
Bulwark™
STG2CHRG3XL
3X-Large
Each for $152.92
Only null left
 
Description

Description

  • The best of two hard-working worlds, this lightweight FR Flex Knit combines the crisp, professional look of an office work shirt and the comfort of your favorite T-shirt.
  • Spandex-blend fabric allows it to stretch freely.
  • Button-down collar keeps you looking crisp and sharp all day.
  • Easy-to-spot CAT 2/2112 label on the front pocket and left sleeve leave no question that you're protected on the job site.
  • Red buttonhole stitching and right sleeve placket leave no doubt you're armed with the very best.
  • Certifications Level 1: NFPA 2112, NFPA 70E
  • Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Arc Flash, Flash Fire
  • Number of Pockets: 1
  • Primary Closure: Button
  • Wash Care: Industrial Laundry - Light Soil
  • Fabric: Body: 6.75 oz. (230 g/m&superscript_2;) Flame Resistant Jersey
  • Blend: Body: 93% Cotton / 7% Spandex Elastane
  • Features: Button down collar, Single chest pocket with button closure, Lightweight, stretch fabric
  • Protection: ATPV Arc Rating: 8.3
  • Country of Origin: Imported, Made in USA Fabric
  • Gender: Male
  • Brand: Bulwark™
Specifications

Specifications

Men's
Charcoal
Long Sleeve
Button
1
8.3
Cotton, Spandex
Collar
Knit
Reusable
Category 2
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'