missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Men's Charcoal Plaid Flame Resistant Flex Knit Button Down Shirt

Catalog No. 17204054
Change view
Click to view available options
Size:
2X-Large (Long)
Large (Long)
Medium (Long)
X-Large (Long)
Small
Medium
Large
X-Large
3X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17204054 Small
17204096 Medium
17204091 Large
17204243 X-Large
17204138 Medium (Long)
17204445 Large (Long)
17204225 X-Large (Long)
17203957 2X-Large (Long)
17204295 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 17204054 Supplier Bulwark™ Supplier No. STG2CH RG S

Men's Charcoal Plaid Flame Resistant Flex Knit Button Down Shirt provides flame-resistant protection in a lightweight blended fabric. The result is unbeatable softness coupled with lasting durability. This casual FR shirt features raglan sleeves as well as rib knit collar and cuffs.

  • The best of two hard-working worlds, this lightweight FR Flex Knit combines the crisp, professional look of an office work shirt and the comfort of your favorite T-shirt.
  • Spandex-blend fabric allows it to stretch freely.
  • Button-down collar keeps you looking crisp and sharp all day.
  • Easy-to-spot CAT 2/2112 label on the front pocket and left sleeve leave no question that you're protected on the job site.
  • Red buttonhole stitching and right sleeve placket leave no doubt you're armed with the very best.
  • Certifications Level 1: NFPA 2112, NFPA 70E
  • Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Arc Flash, Flash Fire
  • Number of Pockets: 1
  • Primary Closure: Button
  • Wash Care: Industrial Laundry - Light Soil
  • Fabric: Body: 6.75 oz. (230 g/m&superscript_2;) Flame Resistant Jersey
  • Blend: Body: 93% Cotton / 7% Spandex Elastane
  • Features: Button down collar, Single chest pocket with button closure, Lightweight, stretch fabric
  • Protection: ATPV Arc Rating: 8.3
  • Country of Origin: Imported, Made in USA Fabric
  • Gender: Male
  • Brand: Bulwark™

Specifications

Size Small
Gender Men's
Material Cotton, Spandex
Color Charcoal
Neck Style Collar
Sleeve Style Long Sleeve
Construction Knit
Closure Type Button
Disposable Reusable
No. of Pockets 1
Hazard Risk Category Category 2
Arc Flash Rating 8.3
Certifications/Compliance NFPA 2112, NFPA 70E
Show More Show Less