missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Membranes for Filtration Unit for SafTest™ Test Kits

Catalog No. MP07DP1000
Change view
Click to view available options
Quantity:
18 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP07DP1000 18 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP07DP1000 Supplier MP Biomedicals™ Supplier No. 07DP1000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Membranes for Filtration Unit for SafTest™ solid sample testing platform.

  • Replacement filters for use with the SafTest sample testing platform.
  • Includes 18 manifolds of 6 filters each.

Specifications

For Use With (Application) Food Quality Testing
Quantity 18 Pk.
Storage Requirements Room temperature