missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Melting and Boiling Point Instruments

Fast and accurate. BUCHI M-560/565 Melting Point Instruments have simple menus and programs that are easy to navigate on the intuitive LCD display, with either manual or automatic, real time video instrument.

Supplier:  BUCHI 11058004

 View more versions of this product

Catalog No. 10987132

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • Fail-safe calibration with automated determination and storage of calibration results
  • Automatic determination of melting and boiling points
  • Highly accurate temperature regulation and measurement
  • Optimized reproducibility with homogeneous packing of the capillary using the Sample Loader M-569
  • Control of almost any type of sample by measuring melting point, boiling point or slip melting point.
  • Double check for difficult samples by comparing manual and automated measurement
  • Observation and replay of the phase transition using color display and video recording
  • PC software included for easy management of methods and results
  • Compliant with Pharmacopeia methods (European, USP, Japanese)
  • Comprehensive IQ/OQ procedures including certified calibration and verification compounds for GLP/GMP laboratories.
Specifications

Specifications

Automatic Melting Point Instrument
100/240 V ±10%
± 0.2°C
370 mm
100/240 V ±10%
200 mm
M-565
4.5 kg
50/60 Hz
150 W
PH. EUR., USP and JP
Color, TFT, 320 x 240, 3.5 in.
Melting and boiling point capillaries
Melting Point instrument, Calibration set; Melting point tubes (100 pcs); Boiling point tubes (10); Sample holder
10°C to 400°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions