missing translation for 'onlineSavingsMsg'
Learn More

Melperone 95.0+%, TCI America™

Catalog No. p-7133330
Change view
Click to view available options
Quantity:
100 mg
Catalog No. Quantity
M2622100MG 100 mg
1 options

Catalog No. M2622100MG

Supplier: TCI America M2622100MG

Only null left

Chemical Identifiers

CAS 3575-80-2
Molecular Formula C16H23ClFNO
Molecular Weight (g/mol) 299.81
MDL Number MFCD00867737
InChI Key MQHYXXIJLKFQGY-UHFFFAOYSA-N
PubChem CID 15387
IUPAC Name 1-[4-(4-fluorophenyl)-4-oxobutyl]-4-methylpiperidin-1-ium chloride
SMILES [Cl-].CC1CC[NH+](CCCC(=O)C2=CC=C(F)C=C2)CC1

Specifications

Melting Point 80°C
Boiling Point 125°C
Quantity 100 mg
Synonym 4′-Fluoro-4-(4-methylpiperidino)butyrophenone
Formula Weight 263.36
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Melperone
TSCA No
Recommended Storage Refrigerator