missing translation for 'onlineSavingsMsg'
Learn More

Meloxicam 98.0+%, TCI America™

Catalog No. p-7132936
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19595G 5 g
M195925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19595G Supplier TCI America Supplier No. M19595G
Only null left

Chemical Identifiers

CAS 71125-38-7
Molecular Formula C14H13N3O4S2
Molecular Weight (g/mol) 351.40
MDL Number MFCD00868752
InChI Key DWMREKMVXIFPFM-ACCUITESSA-N
Synonym meloxicam, mobic, metacam, movalis, meloxicamum, mobicox, mobec, meloxicam mobic, movatec, parocin
PubChem CID 54677470
ChEBI CHEBI:6741
IUPAC Name (3E)-3-{hydroxy[(5-methyl-1,3-thiazol-2-yl)amino]methylidene}-2-methyl-3,4-dihydro-2H-1λ⁶,2-benzothiazine-1,1,4-trione
SMILES CN1\C(=C(\O)NC2=NC=C(C)S2)C(=O)C2=CC=CC=C2S1(=O)=O

Specifications

Melting Point 255°C
Quantity 5 g
UN Number 2811
Formula Weight 351.40
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Meloxicam
RTECSNumber DL0702000
TSCA No
Recommended Storage Refrigerator