missing translation for 'onlineSavingsMsg'
Learn More

Meloxicam Impurity Standard, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022913
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022913 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022913 Supplier Merck Emd Millipore Supplier No. BP630

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

Chemical Name or Material Pharmacopeia & Metrological Institutes Standards
CAS 71125-38-7
Molecular Formula C14H13N3O4S2
Linear Formula C14H13N3O4S2
Quantity 100 mg
Synonym Meloxicam; 4-Hydroxy-2-methyl-N-(5-methyl-2-thiazolyl)-2 H-1,2-benzothiazine-3-carboxamide 1,1-dioxide
Formula Weight 351.40
Grade BP