missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mellitic Acid 98.0+%, TCI America™
$367.22 - $1258.24
Chemical Identifiers
| CAS | 517-60-2 |
|---|---|
| Molecular Formula | C12H6O12 |
| Molecular Weight (g/mol) | 342.168 |
| MDL Number | MFCD00002469 |
| InChI Key | YDSWCNNOKPMOTP-UHFFFAOYSA-N |
| Synonym | Benzenehexacarboxylic Acid |
| PubChem CID | 2334 |
| ChEBI | CHEBI:41089 |
| IUPAC Name | benzene-1,2,3,4,5,6-hexacarboxylic acid |
| SMILES | C1(=C(C(=C(C(=C1C(=O)O)C(=O)O)C(=O)O)C(=O)O)C(=O)O)C(=O)O |
Chemical Identifiers
| 517-60-2 | |
| 342.168 | |
| YDSWCNNOKPMOTP-UHFFFAOYSA-N | |
| 2334 | |
| benzene-1,2,3,4,5,6-hexacarboxylic acid |
| C12H6O12 | |
| MFCD00002469 | |
| Benzenehexacarboxylic Acid | |
| CHEBI:41089 | |
| C1(=C(C(=C(C(=C1C(=O)O)C(=O)O)C(=O)O)C(=O)O)C(=O)O)C(=O)O |
Specifications
| 517-60-2 | |
| White-Yellow | |
| MFCD00002469 | |
| Benzenehexacarboxylic Acid | |
| C1(=C(C(=C(C(=C1C(=O)O)C(=O)O)C(=O)O)C(=O)O)C(=O)O)C(=O)O | |
| 342.168 | |
| CHEBI:41089 | |
| ≥98.0% (T) | |
| Mellitic Acid |
| 288°C | |
| C12H6O12 | |
| 5 g | |
| YDSWCNNOKPMOTP-UHFFFAOYSA-N | |
| benzene-1,2,3,4,5,6-hexacarboxylic acid | |
| 2334 | |
| 342.17 | |
| Crystalline Powder |
Safety and Handling
TSCA : No