Learn More
SignalChem Biotech MARK4, Active 500ug
Catalog No. 502574230
Encompass
Recombinant 1-662 human MARK4 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 502574230 Supplier SignalChem Biotech Supplier No. M4610G500
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant 1-662 human MARK4 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.