missing translation for 'onlineSavingsMsg'
Learn More

Meldrum's Acid (=2,2-Dimethyl-1,3-dioxane-4,6-dione) 98.0+%, TCI America™

Catalog No. M079925G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M079925G 25 g
M0799100G 100 g
M0799500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M079925G Supplier TCI America Supplier No. M079925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2033-24-1
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.126
MDL Number MFCD00006638
InChI Key GXHFUVWIGNLZSC-UHFFFAOYSA-N
Synonym meldrum's acid, isopropylidene malonate, meldrums acid, 1,3-dioxane-4,6-dione, 2,2-dimethyl, sub-isopropyl malonate, 2,2-dimethyl-1,3-dioxan-4,6-dione, cycl-isopropylidene malonate, cyclic isopropylidene malonate, malonic acid cyclic isopropylidene ester, meldrum acid
PubChem CID 16249
IUPAC Name 2,2-dimethyl-1,3-dioxane-4,6-dione
SMILES CC1(OC(=O)CC(=O)O1)C

Specifications

Melting Point 95°C
Quantity 25 g
Formula Weight 144.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Meldrum′s Acid (=2,2-Dimethyl-1,3-dioxane-4,6-dione)
EINECSNumber (5)-6991
RTECSNumber JH0800000
TSCA Yes
Recommended Storage Refrigerator