missing translation for 'onlineSavingsMsg'
Learn More

Meldola's Blue, Spectrum™ Chemical

Catalog No. 18604288
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18604288 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18604288 Supplier Spectrum Chemical Mfg Cor Supplier No. M38075GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

M3807, 7057-57-0, C18H15ClN2O·Cl2Zn

Spectrum™ Chemical Meldola′s Blue

Chemical Identifiers

CAS 7057-57-0
Molecular Formula C18H15Cl3N2OZn
Molecular Weight (g/mol) 447.06
InChI Key YTEJSAFVYHDCSN-UHFFFAOYSA-K
IUPAC Name zinc(2+) 8-(dimethylamino)-10λ⁴-oxa-5-azatetraphen-10-ylium trichloride
SMILES [Cl-].[Cl-].[Cl-].[Zn++].CN(C)C1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C=C3

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Color Dark Green
Quantity 5 g
Grade Reagent
Chemical Name or Material Meldola's Blue