missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Melanotan II

Catalog No. aaj65115mcr
Change view
Click to view available options
Quantity:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65115MCR 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ65115MCR Supplier Thermo Scientific™ Supplier No. J65115MCR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 121062-08-6
Color White
Content And Storage Store at -20°C
Format Solid
Molecular Weight (g/mol) 1024.198
Product Type Melanotan II
Quantity 1mg