missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melamine Monomer 98.0+%, TCI America™
$32.72 - $38.51
Chemical Identifiers
| CAS | 108-78-1 |
|---|---|
| Molecular Formula | C3H6N6 |
| Molecular Weight (g/mol) | 126.12 |
| MDL Number | MFCD00006055 |
| InChI Key | JDSHMPZPIAZGSV-UHFFFAOYSA-N |
| Synonym | melamine, cyanurotriamide, cyanuramide, cyanurotriamine, isomelamine, theoharn, teoharn, triaminotriazine, cyanuric triamide, hicophor pr |
| PubChem CID | 7955 |
| ChEBI | CHEBI:27915 |
| IUPAC Name | 1,3,5-triazine-2,4,6-triamine |
| SMILES | NC1=NC(N)=NC(N)=N1 |
Chemical Identifiers
| 108-78-1 | |
| 126.12 | |
| JDSHMPZPIAZGSV-UHFFFAOYSA-N | |
| 7955 | |
| 1,3,5-triazine-2,4,6-triamine |
| C3H6N6 | |
| MFCD00006055 | |
| melamine, cyanurotriamide, cyanuramide, cyanurotriamine, isomelamine, theoharn, teoharn, triaminotriazine, cyanuric triamide, hicophor pr | |
| CHEBI:27915 | |
| NC1=NC(N)=NC(N)=N1 |
Specifications
| 108-78-1 | |
| C3H6N6 | |
| 25 g | |
| JDSHMPZPIAZGSV-UHFFFAOYSA-N | |
| 1,3,5-triazine-2,4,6-triamine | |
| 7955 | |
| 126.12 | |
| Crystalline Powder |
| White | |
| MFCD00006055 | |
| melamine, cyanurotriamide, cyanuramide, cyanurotriamine, isomelamine, theoharn, teoharn, triaminotriazine, cyanuric triamide, hicophor pr | |
| NC1=NC(N)=NC(N)=N1 | |
| 126.12 | |
| CHEBI:27915 | |
| ≥98.0% (HPLC,T) | |
| Melamine Monomer |
Safety and Handling
EINECSNumber : (5)-1024
RTECSNumber : OS0700000
TSCA : Yes