missing translation for 'onlineSavingsMsg'
Learn More

Melamine Monomer 98.0+%, TCI America™

Catalog No. T0337500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0337500G 500 g
T033725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0337500G Supplier TCI America Supplier No. T0337500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-78-1
Molecular Formula C3H6N6
Molecular Weight (g/mol) 126.12
MDL Number MFCD00006055
InChI Key JDSHMPZPIAZGSV-UHFFFAOYSA-N
Synonym melamine, cyanurotriamide, cyanuramide, cyanurotriamine, isomelamine, theoharn, teoharn, triaminotriazine, cyanuric triamide, hicophor pr
PubChem CID 7955
ChEBI CHEBI:27915
IUPAC Name 1,3,5-triazine-2,4,6-triamine
SMILES NC1=NC(N)=NC(N)=N1

Specifications

Color White
Quantity 500 g
Formula Weight 126.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Melamine Monomer
EINECSNumber (5)-1024
RTECSNumber OS0700000
TSCA Yes