missing translation for 'onlineSavingsMsg'
Learn More

Melamine, 99%

Catalog No. AAA1129536
Change view
Click to view available options
Quantity:
10,000 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1129536 500 g
AAA112950E 2500 g
AAA112950C 10,000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1129536 Supplier Thermo Scientific Chemicals Supplier No. A1129536
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-78-1 | C3H6N6 | 126.12 g/mol

Melamine is used in the production of synthetic resins such as melamine resin with formaldehyde, which is used in high pressure decorative laminates like formica, melamine dinnerware, laminate flooring, and dry erase boards. The presence rich in nitrogen content resulted in fire retardant properties when mixed with resins. It is used to prepare melamine cyanurate by reacting with cyanuric acid. It finds application as superplasticizer for making high-resistance concrete, soundproofing material and in polymeric cleaning products like magic eraser. It is an active component in pigment yellow 150. Further, it is also used as a nitrogen and carbon source for N-doped carbon nanotube.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Melamine is used in the production of synthetic resins such as melamine resin with formaldehyde, which is used in high pressure decorative laminates like formica, melamine dinnerware, laminate flooring, and dry erase boards. The presence rich in nitrogen content resulted in fire retardant properties when mixed with resins. It is used to prepare melamine cyanurate by reacting with cyanuric acid. It finds application as superplasticizer for making high-resistance concrete, soundproofing material and in polymeric cleaning products like magic eraser. It is an active component in pigment yellow 150. Further, it is also used as a nitrogen and carbon source for N-doped carbon nanotube.

Solubility
Slightly soluble in water, hot alcohol, glycerol, glycol and pyridine. Insoluble in ether, benzene and carbon tetrachloride.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 108-78-1
Molecular Formula C3H6N6
Molecular Weight (g/mol) 126.12
MDL Number MFCD00006055
InChI Key JDSHMPZPIAZGSV-UHFFFAOYSA-N
Synonym melamine, cyanurotriamide, cyanuramide, cyanurotriamine, isomelamine, theoharn, teoharn, triaminotriazine, cyanuric triamide, hicophor pr
PubChem CID 7955
ChEBI CHEBI:27915
IUPAC Name 1,3,5-triazine-2,4,6-triamine
SMILES NC1=NC(N)=NC(N)=N1

Specifications

Melting Point ∼350°C (decomposition)
Density 1.573
Quantity 500 g
Beilstein 124341
Merck Index 14,5811
Solubility Information Slightly soluble in water,hot alcohol,glycerol,glycol and pyridine. Insoluble in ether,benzene and carbon tetrachloride.
Formula Weight 126.12
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Melamine
Hazard Category H351-H500
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 203-615-4
RTECSNumber OS0700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only