Learn More
Integra™ Miltex™ MeisterHand™ Tissue Forceps
Catalog No. 12460700
Change view
Click to view available options
Length (English):
4.5 in.
5 in.
5.5 in.
6 in.
8 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
12460700 8 in.
12460697 5 in.
12460695 4.5 in.
12460698 5.5 in.
12460699 6 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 12460700 Supplier Integra™ Miltex™ Supplier No. MH648
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Standard-pattern serrated handles
  • 1 x 2 teeth
  • Non-glare matte finish
  • Stainless-steel
Specifications
Product Type Forceps, Tissue
Material Stainless Steel
Finish Matte
Grade Standard, Surgical
Quantity Each
Teeth Count 1 x 2
Length (English) 8 in.
Length (Metric) 203 mm