Learn More
Integra™ Miltex™ MeisterHand™ Tissue Forceps
Standard-pattern serrated handles
Supplier:  Integra™ Miltex™ MH644
 View more versions of this product
Catalog No. 12-460-698
Only null left
Explore available promotions
Description
Description
  • 1 x 2 teeth
  • Non-glare matte finish
  • Stainless-steel
Specifications
Specifications
5.5 in.
140 mm
Each
Standard, Surgical
Stainless Steel
Forceps, Tissue
Matte
1 x 2
Product Suggestions
Product Suggestions