missing translation for 'onlineSavingsMsg'
Learn More

Integra™ Miltex™ MeisterHand™ Rochester-Carmalt Forceps

Catalog No. 12460706
Change view
Click to view available options
Length (English):
6.5 in.
8 in.
Tip Style:
Curved
Straight
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Tip Style
12460706 8 in. Curved
12460704 6.5 in. Straight
12460705 6.5 in. Curved
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12460706 Supplier Integra™ Miltex™ Supplier No. MH7182

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

  • Stainless steel
  • Non-glare matte finish
  • Longitudinal serrations

Specifications

Product Type Forceps, Rochester-Carmalt
Material Stainless Steel
Finish Non-glare Matte
Quantity Each
Tip Style Curved
Length (English) 8 in.
Length (Metric) 203 mm