Learn More
Integra™ Miltex™ MeisterHand™ Metzenbaum Scissors
Rounded tips for dissection
Supplier: Integra™ Miltex™ MH5182
Description
- Stainless steel
- Cuts tough or delicate tissue
- Non-glare matte finish
- Straight or curved blades
- Blunt points
- SuperCut scissors have a specially designed razor sharp edge that cuts effortlessly through tissue; lower blade has micro fine serrations to help prevent tissue slippage
- Black ring handles
Order Info
SuperCut scissiors designed to cut tissue only; not recommended for use on suture material. Dropping or mishandling will cause the razor-shapr edge to nick or become irregular.
Specifications
| Curved | |
| 17.8 cm | |
| Blunt/Blunt |
| 7 in. | |
| Surgical, Metzenbaum |