missing translation for 'onlineSavingsMsg'
Learn More

Megestrol Acetate 96.0+%, TCI America™

Catalog No. M19491G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19491G 1 g
M19495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19491G Supplier TCI America Supplier No. M19491G

Chemical Identifiers

CAS 595-33-5
Molecular Formula C24H32O4
Molecular Weight (g/mol) 384.516
MDL Number MFCD00056470
InChI Key RQZAXGRLVPAYTJ-GQFGMJRRSA-N
Synonym megestrol acetate, niagestin, megace es, megestryl acetate, megace, megeron, ovaban, ovarid, pallace, magestin
PubChem CID 11683
IUPAC Name [(8R,9S,10R,13S,14S,17R)-17-acetyl-6,10,13-trimethyl-3-oxo-2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate
SMILES CC1=CC2C(CCC3(C2CCC3(C(=O)C)OC(=O)C)C)C4(C1=CC(=O)CC4)C

Specifications

Melting Point 218°C
Color White
Quantity 1 g
Formula Weight 384.52
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Megestrol Acetate
RTECSNumber TU4075000
TSCA No