missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ MegaClean™ Heavy-Duty Cleaner

Catalog No. p-9183000
Change view
Click to view available options
Volume (English):
1 gal.
5 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19035380E 1 gal.
19306970 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19035380E Supplier Micronova™ Supplier No. MC1G
Only null left

A non-flammable, heavy duty cleaner and degreaser that removes grease, adhesives and photo-resist stains from most surfaces. Available in 1 or 5 gallon bottles and a 1 quart spray bottle.

The Micronova MegaClean™ Heavy-Duty Cleaner offers an effective alternative to toxic, flammable, solvent-based cleaners, such as acetone and isopropyl alcohol. Filtered to 0.1 microns, this cleaner is ideal for semiconductor, electronics, aerospace and clean-build operations.

  • Non-flammable, heavy duty cleanroom degreaser
  • Removes grease, adhesives and photo-resist stains from most surfaces
  • Ideal for semiconductor, electronics, aerospace and clean-build operations
  • Filtered to 0.1 microns
  • Available in 1 or 5 gallon refill bottles and a 1 quart spray bottle
  • Biodegradable
  • Water soluble
  • Effective at low concentrations

Specifications

Product Type MegaClean Cleaner
Color-coded No
Handle Type None
Sterility Non-sterile
Description Heavy Duty Detergent, Degreaser
For Use With (Application) Heavy Soils, Adhesive Removal in and Around Cleanroom
For Use With (Equipment) Mops and Buckets or Wipes and Sponge
Volume (English) 1 gal.