missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mefloquine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America M23131G
Specifications
| Mefloquine Hydrochloride | |
| 253°C | |
| C17H17ClF6N2O | |
| 1 g | |
| WESWYMRNZNDGBX-UHFFFAOYNA-N | |
| hydrogen [2,8-bis(trifluoromethyl)quinolin-4-yl](piperidin-2-yl)methanol chloride | |
| 18660697 | |
| ≥98.0% (HPLC) |
| 51773-92-3 | |
| White | |
| MFCD00797519,MFCD00797519 | |
| mefloquine hcl, r-2,8-bis trifluoromethyl quinolin-4-yl s-piperidin-2-yl methanol hydrochloride, r*,s*-2,8-bis-trifluoromethyl-quinolin-4-yl-piperidin-2-yl-methanol hydrochloride, r-2,8-bis trifluoromethyl quinolin-4-yl-2s-piperidin-2-yl methanol;hydrochloride, as-rel-a-2r-2-piperidinyl-2,8-bis trifluoromethyl-4-quinolinemethanol monohydrochloride, +-mefloquine hydrochloride, unii-5y9l3636o3 component, r-2,8-bis trifluoromethyl-4-quinolinyl 2s-2-piperidinyl methanol hydrochloride, r-2,8-bis trifluoromethyl quinolin-4-yl 2s-piperidin-2-yl methanol hydrochloride, s-2,8-bis trifluoromethyl quinolin-4-yl r-piperidin-2-yl methanolhydrochloride | |
| [H+].[Cl-].OC(C1CCCCN1)C1=C2C=CC=C(C2=NC(=C1)C(F)(F)F)C(F)(F)F | |
| 414.78 | |
| 414.78 | |
| Crystalline Powder |
Chemical Identifiers
| 51773-92-3 | |
| 414.78 | |
| WESWYMRNZNDGBX-UHFFFAOYNA-N | |
| 18660697 | |
| [H+].[Cl-].OC(C1CCCCN1)C1=C2C=CC=C(C2=NC(=C1)C(F)(F)F)C(F)(F)F |
| C17H17ClF6N2O | |
| MFCD00797519,MFCD00797519 | |
| mefloquine hcl, r-2,8-bis trifluoromethyl quinolin-4-yl s-piperidin-2-yl methanol hydrochloride, r*,s*-2,8-bis-trifluoromethyl-quinolin-4-yl-piperidin-2-yl-methanol hydrochloride, r-2,8-bis trifluoromethyl quinolin-4-yl-2s-piperidin-2-yl methanol;hydrochloride, as-rel-a-2r-2-piperidinyl-2,8-bis trifluoromethyl-4-quinolinemethanol monohydrochloride, +-mefloquine hydrochloride, unii-5y9l3636o3 component, r-2,8-bis trifluoromethyl-4-quinolinyl 2s-2-piperidinyl methanol hydrochloride, r-2,8-bis trifluoromethyl quinolin-4-yl 2s-piperidin-2-yl methanol hydrochloride, s-2,8-bis trifluoromethyl quinolin-4-yl r-piperidin-2-yl methanolhydrochloride | |
| hydrogen [2,8-bis(trifluoromethyl)quinolin-4-yl](piperidin-2-yl)methanol chloride |
Safety and Handling
RTECSNumber : VC0308000
TSCA : No